Isoquinoline-5-boronic acid hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme:
Isoquinolin-5-ylboronic acid hydrochloride
OR9366-1G
APOSOR9366-1GEA
250
EUR
APOSOR9366-1G
Isoquinoline-5-boronic acid hydrochloride 97%
Isoquinoline-5-boronic acid hydrochloride
Formel:
C₉H₈BNO₂·ClH Molecular Weight: 209.437 g/mol Schmelzpunkt: 204…206 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235064 CAS-Nummer: 1256345-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...