2,5-Dimethyl-3-hexin-2,5-diol 95%

Lieferant: Apollo Scientific

Synonyme: 2,5-Dihydroxy-2,5-dimethyl-3-hexin

OR936618-500G
APOSOR936618-500GEA 74.1 EUR
APOSOR936618-500G
2,5-Dimethyl-3-hexin-2,5-diol 95%
2,5-Dimethyl-3-hexin-2,5-diol
Formel: C₈H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00004468
CAS-Nummer: 142-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR