Phenyl acrylate 95%

Lieferant: Apollo Scientific

OR936700-25G
APOSOR936700-25GEA 116 EUR
APOSOR936700-25G
Phenyl acrylate 95%
Phenyl acrylate
Formel: C₉H₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00048145
CAS-Nummer: 937-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR