(S)-(-)-β-Citronellol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-(-)-Citronellol
OR936732-25G
OR936732-5G
OR936732-100G
APOSOR936732-25GEA
78.3
EUR
APOSOR936732-25G
APOSOR936732-5G
APOSOR936732-100G
(S)-(-)-β-Citronellol 95%
(S)-(-)-β-Citronellol
Formel:
C₁₀H₂₀O Molecular Weight: 156.27 g/mol Siedepunkt: 225…226 °C Dichte: 0.856 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063214 CAS-Nummer: 7540-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...