4-Butylbenzylamin 95%
Lieferant: Apollo Scientific
OR936806-5G
APOSOR936806-5GEA
444
EUR
APOSOR936806-5G
4-Butylbenzylamin 95%
4-Butylbenzylamin
Formel:
C₁₁H₁₇N Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258855 CAS-Nummer: 57802-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...