Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate 96%

Lieferant: Apollo Scientific

OR936808-5G
APOSOR936808-5GEA 78.3 EUR
APOSOR936808-5G
Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate 96%
Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate
Formel: C₁₀H₁₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00209581
CAS-Nummer: 4841-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR