N,4′-Dimethylformanilid 95%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-N-(p-tolyl)formamide

OR936825-1G
APOSOR936825-1GEA 95.7 EUR
APOSOR936825-1G
N,4′-Dimethylformanilid 95%
N,4′-Dimethylformanilid
Formel: C₉H₁₁NO
Molecular Weight: 149.19 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2739-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR