Methylthiosalicylat 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-mercaptobenzoate

OR936843-1G OR936843-25G OR936843-5G OR936843-2.5G OR936843-10G
APOSOR936843-1GEA 26.1 EUR
APOSOR936843-1G APOSOR936843-25G APOSOR936843-5G APOSOR936843-2.5G APOSOR936843-10G
Methylthiosalicylat 95%
Methylthiosalicylat
Formel: C₈H₈O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00060678
CAS-Nummer: 4892-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR