2-Hydroxyethyl-4-hydroxybenzoat 95%

Lieferant: Apollo Scientific

OR936885-100G OR936885-10G OR936885-25G
APOSOR936885-100GEA 734 EUR
APOSOR936885-100G APOSOR936885-10G APOSOR936885-25G
2-Hydroxyethyl-4-hydroxybenzoat 95%
2-Hydroxyethyl-4-hydroxybenzoate
Formel: C₉H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00059079
CAS-Nummer: 2496-90-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR