1,2-Di(2-pyridyl)ethylen 95%

Lieferant: Apollo Scientific

OR936887-1G
APOSOR936887-1GEA 154 EUR
APOSOR936887-1G
1,2-Di(2-pyridyl)ethylen 95%
1,2-Di(2-pyridyl)ethylen
Formel: C₁₂H₁₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00006356
CAS-Nummer: 1437-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR