trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 95%

Lieferant: Apollo Scientific

OR936898-5G
APOSOR936898-5GEA 250 EUR
APOSOR936898-5G
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 95%
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene
Formel: C₈H₁₆O₂Si
Lagertemperatur: Raumtemperatur
MDL: MFCD00008498
CAS-Nummer: 54125-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR