4-Amino-2-methylchinolin 95%
Lieferant: Apollo Scientific
Synonyme:
4-Amino-2-methylchinolin
OR936906-25G
OR936906-5G
APOSOR936906-25GEA
203
EUR
APOSOR936906-25G
APOSOR936906-5G
4-Amino-2-methylchinolin 95%
4-Amino-2-methylchinolin
Formel:
C₁₀H₁₀N₂ Molecular Weight: 158.2 g/mol Siedepunkt: 333 Schmelzpunkt: 168 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006759 CAS-Nummer: 6628-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...