3-Methyl-1,5-pentandiol 95%

Lieferant: Apollo Scientific

OR936911-500G
APOSOR936911-500GEA 66.7 EUR
APOSOR936911-500G
3-Methyl-1,5-pentandiol 95%
3-Methyl-1,5-pentanediol
Formel: C₆H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00039547
CAS-Nummer: 4457-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR