5-Ethoxy-2-mercaptobenzimidazole 97%

Lieferant: Apollo Scientific

OR936925-25G
APOSOR936925-25GEA 0 EUR
APOSOR936925-25G
5-Ethoxy-2-mercaptobenzimidazole 97%
5-Ethoxy-2-mercaptobenzimidazole
Formel: C₉H₁₀N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD01326487
CAS-Nummer: 55489-15-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR