4-(Trimethylsilyl)morpholin 97%
Lieferant: Apollo Scientific
Synonyme:
4-(Trimethylsilyl)morpholine
OR936934-25G
OR936934-100G
APOSOR936934-25GEA
102
EUR
APOSOR936934-25G
APOSOR936934-100G
4-(Trimethylsilyl)morpholin 97%
4-(Trimethylsilyl)morpholin
Formel:
C₇H₁₇NOSi Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010098 CAS-Nummer: 13368-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...