4-(Pentyloxy)phenol 95%

Lieferant: Apollo Scientific

OR936941-1G OR936941-5G
APOSOR936941-1GEA 21.5 EUR
APOSOR936941-1G APOSOR936941-5G
4-(Pentyloxy)phenol 95%
4-(Pentyloxy)phenol
Formel: C₁₁H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00044283
CAS-Nummer: 18979-53-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR