N-(2-Methoxyethyl)Ethylamin 97%

Lieferant: Apollo Scientific

Synonyme: N-Ethyl-2-methoxyethanamine, N-Ethyl-2-methoxyethylamine

OR936987-5G
APOSOR936987-5GEA 151 EUR
APOSOR936987-5G
N-(2-Methoxyethyl)Ethylamin 97%
N-(2-Methoxyethyl)Ethylamin
Formel: C₅H₁₃NO
Molecular Weight: 103,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144876
CAS-Nummer: 34322-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR