4-Chlor-(3-ethoxycarbonyl)phenylboronsäure 96%

Lieferant: Apollo Scientific

OR9370-1G
APOSOR9370-1GEA 84.1 EUR
APOSOR9370-1G
4-Chlor-(3-ethoxycarbonyl)phenylboronsäure 96%
4-Chlor-(3-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Schmelzpunkt: 162…164 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235062
CAS-Nummer: 874219-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR