3-(1,3-Dioxolan-2-yl)benzonitrile 95%
Lieferant: Apollo Scientific
OR937002-1G
APOSOR937002-1GEA
159
EUR
APOSOR937002-1G
3-(1,3-Dioxolan-2-yl)benzonitrile 95%
3-(1,3-Dioxolan-2-yl)benzonitrile
Formel:
C₁₀H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06208939 CAS-Nummer: 153329-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...