Allyl-p-tolyl ether 95%

Lieferant: Apollo Scientific

OR937057-25G OR937057-5G
APOSOR937057-25GEA 0 EUR
APOSOR937057-25G APOSOR937057-5G
Allyl-p-tolyl ether 95%
Allyl-p-tolyl ether
Formel: C₁₀H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00026098
CAS-Nummer: 23431-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR