2,2-Dimethylbuttersäure 98%

Lieferant: Apollo Scientific

OR937096-100G OR937096-500G
APOSOR937096-100GEA 29 EUR
APOSOR937096-100G APOSOR937096-500G
2,2-Dimethylbuttersäure 98%
2,2-Dimethylbuttersäure
Formel: C₆H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00004200
CAS-Nummer: 595-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR