2,2-Dimethylbuttersäure 98%
Lieferant: Apollo Scientific
OR937096-100G
OR937096-500G
APOSOR937096-100GEA
29
EUR
APOSOR937096-100G
APOSOR937096-500G
2,2-Dimethylbuttersäure 98%
2,2-Dimethylbuttersäure
Formel:
C₆H₁₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004200 CAS-Nummer: 595-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...