1-Butyl-2-nitrobenzol 95%

Lieferant: Apollo Scientific

OR937097-5G
APOSOR937097-5GEA 0 EUR
APOSOR937097-5G
1-Butyl-2-nitrobenzol 95%
1-Butyl-2-nitrobenzene
Formel: C₁₀H₁₃NO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 7137-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR