4-Iodbenzylamin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Iodophenyl)methanamine hydrochloride

OR9371-1G OR9371-5G
APOSOR9371-1GEA 21.9 EUR
APOSOR9371-1G APOSOR9371-5G
4-Iodbenzylamin Hydrochlorid
4-Iodbenzylamin Hydrochlorid
Formel: C₇H₉ClIN
Molecular Weight: 269.51 g/mol
Schmelzpunkt: 275…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039324
CAS-Nummer: 59528-27-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR