Ethyl (E)-hex-2-enoat 97%
Lieferant: Apollo Scientific
Synonyme:
trans-2-Hexenoic Acid Ethyl Ester, Ethyl (E)-2-hexenoate, Ethyl (E)-hex-2-enoate
OR937124-25G
APOSOR937124-25GEA
174
EUR
APOSOR937124-25G
Ethyl (E)-hex-2-enoat 97%
Ethyl-trans-2-hexenoate
Formel:
C₈H₁₄O₂ Molecular Weight: 142,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036541 CAS-Nummer: 27829-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...