2,7-Dimethoxy-1,4,5,8-tetrahydronaphthalin 97%
Lieferant: Apollo Scientific
OR937133-5G
APOSOR937133-5GEA
183
EUR
APOSOR937133-5G
2,7-Dimethoxy-1,4,5,8-tetrahydronaphthalin 97%
2,7-Dimethoxy-1,4,5,8-tetrahydronaphthalene
Formel:
C₁₂H₁₆O₂ Molecular Weight: 192,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269695 CAS-Nummer: 1614-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...