2-Methoxybiphenyl 95%

Lieferant: Apollo Scientific

Synonyme: Biphenyl-2-yl methyl ether, 2-Phenylanisole

OR937140-25G
APOSOR937140-25GEA 75.4 EUR
APOSOR937140-25G
2-Methoxybiphenyl 95%
2-Methoxybiphenyl
Formel: C₁₃H₁₂O
Molecular Weight: 184,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008367
CAS-Nummer: 86-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR