4-Ethylbenzonitril 95%
Lieferant: Apollo Scientific
Synonyme:
p-Ethylbenzonitrile
OR937171-25G
APOSOR937171-25GEA
106
EUR
APOSOR937171-25G
4-Ethylbenzonitril 95%
4-Ethylbenzonitril
Formel:
C₉H₉N Molecular Weight: 131.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016383 CAS-Nummer: 25309-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...