4-tert-Butylphenyl acetate 95%
Lieferant: Apollo Scientific
Synonyme:
Acetic Acid 4-tert-Butylphenyl Ester
OR937200-5G
APOSOR937200-5GEA
377
EUR
APOSOR937200-5G
4-tert-Butylphenyl acetate 95%
4-tert-Butylphenyl acetate
Formel:
C₁₂H₁₆O₂ Molecular Weight: 192,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026308 CAS-Nummer: 3056-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...