1,2-Heptandiol 95%

Lieferant: Apollo Scientific

OR937209-10G OR937209-1G OR937209-5G
APOSOR937209-10GEA 683 EUR
APOSOR937209-10G APOSOR937209-1G APOSOR937209-5G
1,2-Heptandiol 95%
1,2-Heptanediol
Formel: C₇H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01861287
CAS-Nummer: 3710-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR