(±)-1-Octen-3-ol 95%

Lieferant: Apollo Scientific

OR937218-500G
APOSOR937218-500GEA 163 EUR
APOSOR937218-500G
(±)-1-Octen-3-ol 95%
(±)-1-Octen-3-ol
Formel: C₈H₁₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00004589
CAS-Nummer: 3391-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR