6-Ethoxychinaldin 95%

Lieferant: Apollo Scientific

Synonyme: 6-Ethoxy-2-methylchinolin

OR937252-5G
APOSOR937252-5GEA 68.3 EUR
APOSOR937252-5G
6-Ethoxychinaldin 95%
6-Ethoxychinaldin
Formel: C₁₂H₁₃NO
Molecular Weight: 187,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023987
CAS-Nummer: 6628-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR