3-Acetylbenzo[b]thiophene 95%

Lieferant: Apollo Scientific

OR937265-1G OR937265-5G
APOSOR937265-1GEA 0 EUR
APOSOR937265-1G APOSOR937265-5G
3-Acetylbenzo[b]thiophene 95%
3-Acetylbenzo[b]thiophene
Formel: C₁₀H₈OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00051638
CAS-Nummer: 1128-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR