Methylsuccinic acid 95%

Lieferant: Apollo Scientific

OR937273-100G
APOSOR937273-100GEA 66.7 EUR
APOSOR937273-100G
Methylsuccinic acid 95%
Methylsuccinic acid
Formel: C₅H₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00002659
CAS-Nummer: 498-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR