3,4-Dichlorthiophen-1,1-dioxid 95%

Lieferant: Apollo Scientific

OR937277-250MG
APOSOR937277-250MGEA 905 EUR
APOSOR937277-250MG
3,4-Dichlorthiophen-1,1-dioxid 95%
3,4-Dichlorthiophen-1,1-dioxid
Formel: C₄H₂Cl₂O₂S
Molecular Weight: 185.02 g/mol
Schmelzpunkt: 108 °C (Zers.)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 52819-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR