2,2-Dimethyl-4-pentensäure 95%

Lieferant: Apollo Scientific

OR937287-5G
APOSOR937287-5GEA 140 EUR
APOSOR937287-5G
2,2-Dimethyl-4-pentensäure 95%
2,2-Dimethyl-4-pentenoic acid
Formel: C₇H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00012304
CAS-Nummer: 16386-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR