trans-Stilbene oxide 98%

Lieferant: Apollo Scientific
Achtung

OR9373-5G
APOSOR9373-5GEA 81.2 EUR
APOSOR9373-5G
trans-Stilbene oxide 98%
trans-Stilbene oxide
Formel: C₁₄H₁₂O
Schmelzpunkt: 65…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064311
CAS-Nummer: 1439-07-2
EINECS: 215-877-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR