1,4-Dimethoxy-2-nitrobenzol 95%
Lieferant: Apollo Scientific
Synonyme:
2-Nitrohydroquinone dimethyl ether, Nitrodimethylhydroquinone
OR937358-25G
APOSOR937358-25GEA
171
EUR
APOSOR937358-25G
1,4-Dimethoxy-2-nitrobenzol 95%
1,4-Dimethoxy-2-nitrobenzene
Formel:
C₈H₉NO₄ Molecular Weight: 183.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024211 CAS-Nummer: 89-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...