3-(Dimethylcarbamoyl)-5-nitrobenzeneboronic acid 96%

Lieferant: Apollo Scientific

OR9374-1G
APOSOR9374-1GEA 205 EUR
APOSOR9374-1G
3-(Dimethylcarbamoyl)-5-nitrobenzeneboronic acid 96%
3-(Dimethylcarbamoyl)-5-nitrobenzeneboronic acid
Formel: C₉H₁₁BN₂O₅
Molecular Weight: 238,005 g/mol
Schmelzpunkt: 160…162 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235061
CAS-Nummer: 874219-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR