2-Butyl-3-methylpyrazin 95%

Lieferant: Apollo Scientific

OR937428-1G
APOSOR937428-1GEA 0 EUR
APOSOR937428-1G
2-Butyl-3-methylpyrazin 95%
2-Butyl-3-methylpyrazine
Formel: C₉H₁₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00038031
CAS-Nummer: 15987-00-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR