1-(1-Pyrrolidinyl)ethanon 95%

Lieferant: Apollo Scientific

Synonyme: 1-(Pyrrolidin-1-yl)ethanone

OR937467-5G
APOSOR937467-5GEA 120 EUR
APOSOR937467-5G
1-(1-Pyrrolidinyl)ethanon 95%
1-Acetylpyrrolidine
Formel: C₆H₁₁NO
Molecular Weight: 113,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014096
CAS-Nummer: 4030-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR