2-(Ethylmethylamino)pyridine 95%

Lieferant: Apollo Scientific

OR937540-1G OR937540-250MG
APOSOR937540-1GEA 0 EUR
APOSOR937540-1G APOSOR937540-250MG
2-(Ethylmethylamino)pyridine 95%
2-(Ethylmethylamino)pyridine
Formel: C₈H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04038419
CAS-Nummer: 77200-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR