6-Chloro-n4,n4-dimethylpyrimidine-2,4-diamine 95%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-N4,N4-dimethylpyrimidine-2,4-diamine
OR937559-1G
OR937559-5G
APOSOR937559-1GEA
105
EUR
APOSOR937559-1G
APOSOR937559-5G
6-Chloro-n4,n4-dimethylpyrimidine-2,4-diamine 95%
6-Chloro-n4,n4-dimethylpyrimidine-2,4-diamine
Formel:
C₆H₉ClN₄ Molecular Weight: 172.615 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13188630 CAS-Nummer: 1007-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...