2-(2-Chlorethoxy)tetrahydropyran 96%

Lieferant: Apollo Scientific

OR937581-25G OR937581-5G
APOSOR937581-25GEA 129 EUR
APOSOR937581-25G APOSOR937581-5G
2-(2-Chlorethoxy)tetrahydropyran 96%
Formel: C₇H₁₃ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00142761
CAS-Nummer: 5631-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR