2,3-Dicyan-5-methylpyrazin 98%
Lieferant: Apollo Scientific
Synonyme:
5-Methylpyrazine-2,3-dicarbonitrile, 5-Methyl-2,3-pyrazinedicarbonitrile
OR937582-5G
APOSOR937582-5GEA
89.9
EUR
APOSOR937582-5G
2,3-Dicyan-5-methylpyrazin 98%
2,3-Dicyano-5-methylpyrazine
Formel:
C₇H₄N₄ Molecular Weight: 144,1300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191635 CAS-Nummer: 52197-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...