Vinyl-2-chlorobenzoate 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chlorobenzoic Acid Vinyl Ester

OR937613-5G
APOSOR937613-5GEA 464 EUR
APOSOR937613-5G
Vinyl-2-chlorobenzoate 95%
Vinyl-2-chlorobenzoate
Formel: C₉H₇ClO₂
Molecular Weight: 182,60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059479
CAS-Nummer: 15721-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR