Acrylsäure-Isoamylester 95%

Lieferant: Apollo Scientific

OR937646-25G OR937646-5G
APOSOR937646-25GEA 502 EUR
APOSOR937646-25G APOSOR937646-5G
Acrylsäure-Isoamylester 95%
Isoamyl acrylate
Formel: C₈H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00053725
CAS-Nummer: 4245-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR