(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol 95%

Lieferant: Apollo Scientific

Synonyme: (1S)-1-(2-chlorophenyl)ethane-1,2-diol

OR937676-5G
APOSOR937676-5GEA 74.1 EUR
APOSOR937676-5G
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol 95%
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol
Formel: C₈H₉ClO₂
Molecular Weight: 172.608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863241
CAS-Nummer: 133082-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR