5-Norbornene-2,2-dimethanol 95%

Lieferant: Apollo Scientific

Synonyme: 2,2-Bis(hydroxymethyl)-5-norbornene, Bicyclo[2.2.1]hept-5-ene-2,2-dimethanol

OR937742-1G OR937742-5G
APOSOR937742-1GEA 78.3 EUR
APOSOR937742-1G APOSOR937742-5G
5-Norbornene-2,2-dimethanol 95%
5-Norbornene-2,2-dimethanol
Formel: C₉H₁₄O₂
Molecular Weight: 154.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00167595
CAS-Nummer: 6707-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR