Trimethylsilylmethyl acetate 95%

Lieferant: Apollo Scientific

Synonyme: (Trimethylsilyl)methyl acetate, Acetic Acid Trimethylsilylmethyl Ester

OR937767-25G
APOSOR937767-25GEA 72.5 EUR
APOSOR937767-25G
Trimethylsilylmethyl acetate 95%
Trimethylsilylmethyl acetate
Formel: C₆H₁₄O₂Si
Molecular Weight: 146.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009650
CAS-Nummer: 2917-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR